missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGC1a Mouse anti-Human, Unconjugated, Clone: 1C1B2, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | PGC1a |
---|---|
Clone | 1C1B2 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16838185
|
Proteintech
66369-1-IG-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16828185
|
Proteintech
66369-1-IG-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PGC1a antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human PGC1a. This antibody recognizes human, mouse, rat antigen. The PGC1a antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
PGC1a | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
10891 | |
Primary |
1C1B2 | |
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG1 |