missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ PFA Wide Neck Reagent Bottle
Click to view available options
Capacity (Metric):
1000 mL
Description
- PerFluoroAlkoxy (PFA) construction
- Narrow neck
- Autoclavable
- Easy to clean
- Withstands temperatures from -200 to 260°C
- Includes thread S28 and screw cap
Specifications
Specifications
Capacity (Metric) | 1000 mL |
For Use With (Application) | For Long-Term Storage of Solvents and Standards |
Material | PFA |
Temperature Range (Metric) | –200 to 260°C |
Autoclavable | Autoclavable |
Product Type | Wide Mouth Bottle |
Closure Type | Screw Cap |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'