Learn More
Cole-Parmer™ PFA Threaded Plug
Threaded fittings offer more permanent installation
29.35 € - 52.10 €
Specifications
Type Plug
Color Clear
Port Type Male NPT Fitting
Sterilization Method Ethylene Oxide (ETO) Only
Overall Length (English) 27/32 in.
View More Specs
Products 2
Product Code Brand Thread Size Price Quantity & Availability  
Product Code Brand Thread Size Price Quantity & Availability  
11775758
View Documents
Cole-Parmer™
06546-40
1/2 in.
52.10 €
Each
Estimated Shipment: 24-02-2025
to see stock.
Add to basket
11755758
View Documents
Cole-Parmer™
06546-39
3/8 in.
29.35 €
Each
Estimated Shipment: 24-02-2025
to see stock.
Add to basket
Description
Description
  • Chemically-inert PFA handles most aggressive chemicals
  • Threaded reducing bushing
  • Choose the size and material that meets your application needs
  • Use with our rigid tubing for your high-pressure applications
Specifications
Specifications
Plug
Male NPT Fitting
27/32 in.
Clear
Ethylene Oxide (ETO) Only
PFA