missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether, puriss., Meets analytical specification of DAB, Honeywell™
BIOPHARMA

Puriss., low boiling point hydrogen treated naphtha, meets analytical specification of DAB, bpmin. 75% 40-60 °C (min. 75%)

70.45 € - 1116.82 €

Chemical Identifiers

CAS 101316-46-5
MDL Number MFCD00081849
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15632680
BIOPHARMA
View Documents
Honeywell Riedel-de Haën
24587-1L
1 L
70.45 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15642680
BIOPHARMA
View Documents
Honeywell Riedel-de Haën
24587-2.5L
2.5 L
144.55 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15682680
BIOPHARMA
View Documents
Honeywell Riedel-de Haën
24587-5L
5 L
286.53 €
5 L
Please to purchase this item. Need a web account? Register with us today!
15652680
BIOPHARMA
View Documents
Honeywell Riedel-de Haën
24587-25L
25 L
1116.82 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications

Chemical Identifiers

101316-46-5
MFCD00081849

Specifications

101316-46-5
−40°C (−40°F)
1268
Petroleum Ether, Meets analytical specification of DAB
0.642 to 0.656g/mL at 20°C
MFCD00081849
puriss.
Videos
Safety and Handling

Safety and Handling

P210-P273-P301 + P310-P331-P391-P403 + P235

H225-H304-H336-H411

Danger

missing translation for 'einecsNumber' : 309-852-0

Product Identifier
  • Petroleum ether

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Hydrocarbons, C6, pentane

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.