missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum Ether, puriss., Honeywell™
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
Specifications
Density | 0.660 to 0.680g/mL at 20°C |
Quantity | 2.5 L |
Flash Point | <−20°C (<−4°F) |
UN Number | 1268 |
Grade | puriss. |
Chemical Name or Material | Petroleum Ether |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction