missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether, puriss., Honeywell™

Product Code. 15612680
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Puriss., high boiling, bp 60-80 °C

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101316-46-5
MDL Number MFCD00081849

Specifications

Density 0.660 to 0.680g/mL at 20°C
Quantity 5 L
Flash Point <−20°C (<−4°F)
UN Number 1268
Grade puriss.
Chemical Name or Material Petroleum Ether
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.