Learn More
Peroxiredoxin 2 Mouse anti-Human, CoraLite 488, Clone: 3H6C4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-60202-100UL
This item is not returnable.
View return policy
Description
The peroxiredoxin 2 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human peroxiredoxin 2. This antibody recognizes human, mouse antigen. The peroxiredoxin 2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Peroxiredoxin 2 | |
Monoclonal | |
2000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse |
Immunofluorescence | |
3H6C4 | |
CoraLite 488 | |
100 μL | |
7001 | |
IgG1 |