missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Periodic Acid (White to Pale-Yellow Crystals or White Powder), Fisher BioReagents
Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly Bottle
Description
Periodic Acid is used to decompose organic compounds.
Specifications
Specifications
pH | 1 |
Color | White |
Melting Point | 122°C |
Density | 1.4 g/cm3 |
Heavy Metals (as Pb) | 0.005% max. |
Identification | Pass Test |
Packaging | Poly Bottle |
Assay Percent Range | ≥99 % |
Quantity | 100 g |
Linear Formula | H5IO6 |
Show More |
Safety and Handling
Product Identifier
- Periodic acid
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Oxidising solid Category 2
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H290-May be corrosive to metals.
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'