Learn More
Perfluoropentanoic acid, 97%, Thermo Scientific Chemicals
68.60 € - 232.00 €
Chemical Identifiers
CAS | 2706-90-3 |
---|---|
Molecular Formula | C5HF9O2 |
Molecular Weight (g/mol) | 264.05 |
MDL Number | MFCD00040211 |
InChI Key | CXZGQIAOTKWCDB-UHFFFAOYSA-N |
Synonym | perfluoropentanoic acid, perfluorovaleric acid, nonafluoropentanoic acid, nonafluorovaleric acid, pentanoic acid, nonafluoro, nonafluoro-1-pentanoic acid, valeric acid, nonafluoro, n-perfluoropentanoic acid, perfluoro-n-pentanoic acid, 2,2,3,3,4,4,5,5,5-nonafluoro-pentanoic acid |
PubChem CID | 75921 |
ChEBI | CHEBI:83491 |
IUPAC Name | 2,2,3,3,4,4,5,5,5-nonafluoropentanoic acid |
SMILES | OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
Description
It is used in the analysis of perfluorooctanesulfonate and related fluorochemicals in water and biological tissue samples by liquid chromatography-ion trap mass spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the analysis of perfluorooctanesulfonate and related fluorochemicals in water and biological tissue samples by liquid chromatography-ion trap mass spectrometry.
Solubility
Partly soluble in water.
Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
2706-90-3 | |
264.05 | |
CXZGQIAOTKWCDB-UHFFFAOYSA-N | |
75921 | |
2,2,3,3,4,4,5,5,5-nonafluoropentanoic acid |
C5HF9O2 | |
MFCD00040211 | |
perfluoropentanoic acid, perfluorovaleric acid, nonafluoropentanoic acid, nonafluorovaleric acid, pentanoic acid, nonafluoro, nonafluoro-1-pentanoic acid, valeric acid, nonafluoro, n-perfluoropentanoic acid, perfluoro-n-pentanoic acid, 2,2,3,3,4,4,5,5,5-nonafluoro-pentanoic acid | |
CHEBI:83491 | |
OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
Specifications
2706-90-3 | |
140°C | |
1.294 | |
UN3265 | |
perfluoropentanoic acid, perfluorovaleric acid, nonafluoropentanoic acid, nonafluorovaleric acid, pentanoic acid, nonafluoro, nonafluoro-1-pentanoic acid, valeric acid, nonafluoro, n-perfluoropentanoic acid, perfluoro-n-pentanoic acid, 2,2,3,3,4,4,5,5,5-nonafluoro-pentanoic acid | |
CXZGQIAOTKWCDB-UHFFFAOYSA-N | |
2,2,3,3,4,4,5,5,5-nonafluoropentanoic acid | |
75921 | |
264.06 | |
Perfluoropentanoic acid |
1.713 | |
C5HF9O2 | |
MFCD00040211 | |
1800087 | |
Partly soluble in water. | |
OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F | |
264.05 | |
CHEBI:83491 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 220-300-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Perfluoropentanoic acid
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Perfluoropentanoic acid
RUO – Research Use Only