Learn More
Perfluorooctanoyl chloride, 97%, Thermo Scientific™
47.84 € - 171.00 €
Chemical Identifiers
CAS | 335-64-8 |
---|---|
Molecular Formula | C8ClF15O |
Molecular Weight (g/mol) | 432.51 |
MDL Number | MFCD00013654 |
InChI Key | AQQBRCXWZZAFOK-UHFFFAOYSA-N |
Synonym | perfluorooctanoyl chloride, pentadecafluorooctanoyl chloride, pentadecafluorooctyl chloride, octanoyl chloride, pentadecafluoro, perfluorooctanoylchloride, perfluorooctanyl chloride, perflurooctanoyl chloride, acmc-1cmee, perfluorocaprylic chloride, n-perfluorooctanoyl chloride |
PubChem CID | 78978 |
SMILES | FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(Cl)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11367326
|
Thermo Scientific Alfa Aesar
L09294.06 |
5 g | |||||||
11377326
|
Thermo Scientific Alfa Aesar
L09294.14 |
25 g |
171.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsPerfluorooctanoyl chloride is derivatizated and used in the determination of Amphetamine and Methamphetamine in Blood. It can be used in agrochemical, pharmaceutical and dyestuff field .
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store away from oxidizing agents and moisture. Store under dry inert gas. Incompatible with strong oxidizing agents, strong bases, alcohols.
Chemical Identifiers
335-64-8 | |
432.51 | |
AQQBRCXWZZAFOK-UHFFFAOYSA-N | |
78978 |
C8ClF15O | |
MFCD00013654 | |
perfluorooctanoyl chloride, pentadecafluorooctanoyl chloride, pentadecafluorooctyl chloride, octanoyl chloride, pentadecafluoro, perfluorooctanoylchloride, perfluorooctanyl chloride, perflurooctanoyl chloride, acmc-1cmee, perfluorocaprylic chloride, n-perfluorooctanoyl chloride | |
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(Cl)=O |
Specifications
335-64-8 | |
130°C to 132°C | |
1.3045 | |
UN3265 | |
Moisture sensitive | |
Reacts with water. | |
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(Cl)=O | |
432.51 | |
432.52 | |
Perfluorooctanoyl chloride |
1.744 | |
C8ClF15O | |
MFCD00013654 | |
1809677 | |
perfluorooctanoyl chloride, pentadecafluorooctanoyl chloride, pentadecafluorooctyl chloride, octanoyl chloride, pentadecafluoro, perfluorooctanoylchloride, perfluorooctanyl chloride, perflurooctanoyl chloride, acmc-1cmee, perfluorocaprylic chloride, n-perfluorooctanoyl chloride | |
AQQBRCXWZZAFOK-UHFFFAOYSA-N | |
2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoyl chloride | |
78978 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 206-394-2
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen