Learn More
(Perfluorocyclohexyl)methyl acrylate, 96%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
88.44 € - 387.00 €
Chemical Identifiers
CAS | 40677-94-9 |
---|---|
Molecular Formula | C10H5F11O2 |
Molecular Weight (g/mol) | 366.13 |
MDL Number | MFCD00080614 |
InChI Key | OZGWOALFBHODRB-UHFFFAOYSA-N |
Synonym | perfluorocyclohexylmethyl acrylate, perfluorocyclohexyl methyl acrylate, undecafluorocyclohexyl methyl prop-2-enoate, 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl methyl prop-2-enoate, acrylic acid 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexylmethyl ester, 1,2,2,3,3,4,4,5,5,6,6-undecakis fluoranyl cyclohexyl methyl prop-2-enoate, 2-propenoic acid 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl methyl ester |
PubChem CID | 2776049 |
IUPAC Name | (1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl)methyl prop-2-enoate |
SMILES | C=CC(=O)OCC1(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(F)F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11470071
|
Thermo Scientific Alfa Aesar
044793.06 |
5 g |
88.44 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480071
|
Thermo Scientific Alfa Aesar
044793.14 |
25 g |
387.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
(Perfluorocyclohexyl)methyl acrylate is used in pharmaceutical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
40677-94-9 | |
366.13 | |
OZGWOALFBHODRB-UHFFFAOYSA-N | |
2776049 | |
C=CC(=O)OCC1(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(F)F)F |
C10H5F11O2 | |
MFCD00080614 | |
perfluorocyclohexylmethyl acrylate, perfluorocyclohexyl methyl acrylate, undecafluorocyclohexyl methyl prop-2-enoate, 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl methyl prop-2-enoate, acrylic acid 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexylmethyl ester, 1,2,2,3,3,4,4,5,5,6,6-undecakis fluoranyl cyclohexyl methyl prop-2-enoate, 2-propenoic acid 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl methyl ester | |
(1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl)methyl prop-2-enoate |
Specifications
40677-94-9 | |
C10H5F11O2 | |
Air sensitive | |
Sparingly soluble in water. | |
C=CC(=O)OCC1(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(F)F)F | |
366.13 | |
366.13 | |
Liquid |
Colorless | |
MFCD00080614 | |
perfluorocyclohexylmethyl acrylate, perfluorocyclohexyl methyl acrylate, undecafluorocyclohexyl methyl prop-2-enoate, 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl methyl prop-2-enoate, acrylic acid 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexylmethyl ester, 1,2,2,3,3,4,4,5,5,6,6-undecakis fluoranyl cyclohexyl methyl prop-2-enoate, 2-propenoic acid 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl methyl ester | |
OZGWOALFBHODRB-UHFFFAOYSA-N | |
(1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl)methyl prop-2-enoate | |
2776049 | |
96% | |
(Perfluorocyclohexyl)methyl acrylate, Stabilized with 100ppm 4-methoxyphenol |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.