Learn More
Perfluoro(decahydronaphthalene), cis + trans, 95%, Thermo Scientific Chemicals
47.40 € - 654.00 €
Chemical Identifiers
CAS | 306-94-5 |
---|---|
Molecular Formula | C10F18 |
Molecular Weight (g/mol) | 462.081 |
MDL Number | MFCD00010626 |
InChI Key | UWEYRJFJVCLAGH-UHFFFAOYSA-N |
Synonym | perfluorodecalin, perflunafene, octadecafluorodecahydronaphthalene, cis-perfluorodecalin, trans-perfluorodecalin, perfluorodecahydronaphthalene, naphthalene, octadecafluorodecahydro, f-dc, pp 5, perflunafenum latin |
PubChem CID | 9386 |
ChEBI | CHEBI:38848 |
IUPAC Name | 1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8,8a-octadecafluoronaphthalene |
SMILES | C12(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(C(C(C(C2(F)F)(F)F)(F)F)(F)F)F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11413837
|
Thermo Scientific Alfa Aesar
A18288.14 |
25 g |
47.40 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11423837
|
Thermo Scientific Alfa Aesar
A18288.22 |
100 g |
140.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11433837
|
Thermo Scientific Alfa Aesar
A18288.36 |
500 g |
654.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Perfluoro(decahydronaphthalene) is used to dissolve gases. Due to its gas carrying capacity, it is utilized to increase oxygen delivery during cell culture. It acts as an ingredient in fluosol which is an artificial blood product. It is also used to dissolve Teflon.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPerfluoro(decahydronaphthalene) is used to dissolve gases. Due to its gas carrying capacity, it is utilized to increase oxygen delivery during cell culture. It acts as an ingredient in fluosol which is an artificial blood product. It is also used to dissolve Teflon.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
306-94-5 | |
462.081 | |
UWEYRJFJVCLAGH-UHFFFAOYSA-N | |
9386 | |
1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8,8a-octadecafluoronaphthalene |
C10F18 | |
MFCD00010626 | |
perfluorodecalin, perflunafene, octadecafluorodecahydronaphthalene, cis-perfluorodecalin, trans-perfluorodecalin, perfluorodecahydronaphthalene, naphthalene, octadecafluorodecahydro, f-dc, pp 5, perflunafenum latin | |
CHEBI:38848 | |
C12(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(C(C(C(C2(F)F)(F)F)(F)F)(F)F)F)F |
Specifications
306-94-5 | |
1.94 | |
C10F18 | |
MFCD00010626 | |
14,4183 | |
Insoluble in water. | |
C12(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(C(C(C(C2(F)F)(F)F)(F)F)(F)F)F)F | |
462.081 | |
CHEBI:38848 | |
95% |
-10°C to -7°C | |
141°C to 142°C | |
1.3145 | |
2067113 | |
perfluorodecalin, perflunafene, octadecafluorodecahydronaphthalene, cis-perfluorodecalin, trans-perfluorodecalin, perfluorodecahydronaphthalene, naphthalene, octadecafluorodecahydro, f-dc, pp 5, perflunafenum latin | |
UWEYRJFJVCLAGH-UHFFFAOYSA-N | |
1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8,8a-octadecafluoronaphthalene | |
9386 | |
462.1 | |
Perfluoro(decahydronaphthalene), cis + trans |
Safety and Handling
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: III
missing translation for 'einecsNumber' : 206-192-4
missing translation for 'rtecsNumber' : QJ3175000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only