missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perchloric Acid 60% d=1.53, Certified AR for Analysis, Fisher Chemical™
99.90 € - 1740.00 €
Chemical Identifiers
CAS | 7601-90-3 |
---|---|
Molecular Formula | ClHO4 |
Molecular Weight (g/mol) | 100.45 |
MDL Number | MFCD00011325 |
InChI Key | VLTRZXGMWDSKGL-UHFFFAOYSA-N |
Synonym | unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent |
PubChem CID | 24247 |
ChEBI | CHEBI:29221 |
IUPAC Name | perchloric acid |
SMILES | O[Cl](=O)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10636652
|
Fisher Chemical
P/1240/PB08 |
500 mL | Plastic Coated Glass Bottle |
99.90 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15908916
|
Fisher Chemical
P/1240/PB15 |
1 L | Amber Glass Bottle |
132.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10646652
|
Fisher Chemical
P/1240/PB17 |
2.5 L | Plastic Coated Glass Bottle |
200.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12606757
|
Fisher Chemical
P/1240/25 |
25 L | Amber Glass Bottle |
1740.00 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
7601-90-3 | |
100.45 | |
VLTRZXGMWDSKGL-UHFFFAOYSA-N | |
24247 | |
perchloric acid |
ClHO4 | |
MFCD00011325 | |
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent | |
CHEBI:29221 | |
O[Cl](=O)(=O)=O |
Specifications
0.1 | |
7601-90-3 | |
203°C | |
ClHO4 | |
Liquid | |
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent | |
O[Cl](=O)(=O)=O | |
100.45 | |
CHEBI:29221 |
Colorless | |
-18°C | |
6.8mmHg 25 | |
3.5 mPaS at 20°C | |
MFCD00011325 | |
VLTRZXGMWDSKGL-UHFFFAOYSA-N | |
perchloric acid | |
24247 | |
100.46g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Perchloric acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Oxidising liquid Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
- H290-May be corrosive to metals.
- H271-May cause fire or explosion; strong oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title