missing translation for 'onlineSavingsMsg'
Learn More

Pentane CHROMASOLV™, for HPLC, ≥99.0%, Honeywell Riedel-de Haën™

CHROMASOLV™, for HPLC,≥99.0%

129.49 € - 657.11 €

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC
View More Specs

Alert:

Repeated exposure may cause skin dryness or cracking.


Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15654720
View Documents
Honeywell-Chromasolv
34956-1L
1 L
129.49 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15664720
View Documents
Honeywell-Chromasolv
34956-2.5L
2.5 L
212.06 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15684720
View Documents
Honeywell-Chromasolv
34956-6X1L
6 x 1 L
609.39 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15674720
View Documents
Honeywell-Chromasolv
34956-4X2.5L
4 x 2.5 L
657.11 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
Specifications

Chemical Identifiers

109-66-0
72.15
OFBQJSOFQDEBGM-UHFFFAOYSA-N
CHEBI:37830
CCCCC
C5H12
MFCD00009498
8003
pentane

Specifications

-130°C
0.626g/mL at 25°C
n20/D 1.358
CH3(CH2)3CH3
969132
OFBQJSOFQDEBGM-UHFFFAOYSA-N
pentane
8003
72.15g/mol
CHROMASOLV™
109-66-0
35°C to 36°C
C5H12
MFCD00009498
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
CCCCC
72.15
CHEBI:37830
≥99%
Pentane, for HPLC
Videos
Safety and Handling

Safety and Handling

P210-P261-P273-P301 + P310-P331

H225-H304-H336-H411

missing translation for 'einecsNumber' : 203-692-4

missing translation for 'rtecsNumber' : RZ9450000

Product Identifier
  • Pentane

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.