missing translation for 'onlineSavingsMsg'
Learn More

Pentane, ≥99.0%, Honeywell Riedel-de Haen™

CHROMASOLV™, for pesticide residue analysis,≥99.0%

164.69 € - 372.07 €

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15697040
View Documents
Honeywell-Chromasolv
76869-1L
1 L Poly Bottle
164.69 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15607050
View Documents
Honeywell-Chromasolv
76869-2.5L
2.5 L Poly Bottle
372.07 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications

Chemical Identifiers

109-66-0
72.15
OFBQJSOFQDEBGM-UHFFFAOYSA-N
8003
pentane
C5H12
MFCD00009498
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
CHEBI:37830
CCCCC

Specifications

5.7 (at 22°C)
109-66-0
1393°C (1013 hPa)
500°C
CH3(CH2)3CH3
UN1265
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
OFBQJSOFQDEBGM-UHFFFAOYSA-N
pentane
8003
72.15g/mol
Pentane
750°C
0.626g/mL at 25°C
8.3%
C5H12
MFCD00009498
969132
946g/L at 25°C
CCCCC
72.15
CHEBI:37830
≥99.0%
Videos
Safety and Handling

Safety and Handling

P210-P261-P273-P301 + P310-P331

missing translation for 'dotInformation' : class 3 - PG 2 - Pentanes

missing translation for 'rtecsNumber' : 203-692-4

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.