missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Pentafluoroethyl)trimethylsilane, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
(Pentafluoroethyl)trimethylsilane acts as a silicon-based fluorinating reagent for trifluoromethylation of carbonyls and ethylene diamines. It is also employed as a precursor for metal-mediated trifluoromethylation of aromatic, heteroaromatic and vinyl substrates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(Pentafluoroethyl)trimethylsilane acts as a silicon-based fluorinating reagent for trifluoromethylation of carbonyls and ethylene diamines. It is also employed as a precursor for metal-mediated trifluoromethylation of aromatic, heteroaromatic and vinyl substrates.
Solubility
Miscible with tetrahydrofuran.
Notes
Moisture sensitive. Incompatible with oxidizing agents.
(Pentafluoroethyl)trimethylsilane acts as a silicon-based fluorinating reagent for trifluoromethylation of carbonyls and ethylene diamines. It is also employed as a precursor for metal-mediated trifluoromethylation of aromatic, heteroaromatic and vinyl substrates.
Solubility
Miscible with tetrahydrofuran.
Notes
Moisture sensitive. Incompatible with oxidizing agents.
Specifications
Specifications
Quantity | 25 g |
UN Number | UN1993 |
Solubility Information | Miscible with tetrahydrofuran. |
Formula Weight | 192.21 |
Percent Purity | 97% |
Refractive Index | 1.327 |
Sensitivity | Moisture sensitive |
Chemical Name or Material | (Pentafluoroethyl)trimethylsilane |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'