missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Electrically insulated headband (dielectric) version available
- Large space inside cup helps reduce moisture and heat build-up
- Sealing rings filled with a unique combination of liquid and foam
- Easy-to-replace cushions and inserts helps keep them hygienically clean
- Soft, wide cusions help reduce pressure around the ears and improves comfort and wearability

Specifications
Specifications
Color | Green |
Certifications/Compliance | CE Approved |
For Use With (Application) | Suitable Choice for Environments With Substantial Industrial Noise or Construction Machinery, Like Airports and Agricultural Work |
Plug Shape | Oval |
Product Type | Ear Muffs Optime II |
Dielectric | Yes |
Material | Stainless Steel |
Style | Headband |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ PELTOR™ Optime™ II Ear Muffs >
Spot an opportunity for improvement?Share a Content Correction