missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ PELTOR™ Optime™ I Ear Muffs
Click to view available options
Model:
H510P3EB-405-GU
Style:
Hard Hat
Description
- Low-profile design which minimizes obtrusiveness
- Large space inside cup helps reduce moisture and heat build-up
- Easy-to-replace cushions and inserts helps keep them hygienically clean
- Soft, wide cushions help reduce pressure around the ears and improve comfort and wearability
Specifications
Specifications
Color | Yellow |
Certifications/Compliance | CE Approved |
For Use With (Application) | Provide Protection Against Noise Arising From a Wide Range of Applications in the Workplace and Leisure Activities |
Model | H510P3EB-405-GU |
Noise Reduction Rating (NRR) | 26 dB |
Plug Shape | Oval |
Product Type | I Ear Muffs |
Material | Stainless Steel |
Style | Hard Hat |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction