Learn More
Thermo Scientific™ PEEKsil™ Capillary Tubing
Get excellent chemical compatibility and very low carryover using the highly inert Thermo Scientific™ PEEKsil™ Capillary Tubing.
Brand:  Thermo Scientific™ 60182-506
 View more versions of this product
Product Code. 10690426
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Get excellent chemical compatibility and very low carryover to improve liquid chromatography (LC) performance using the highly inert Thermo Scientific™ PEEKsil™ Capillary Tubing. Made of precision-bore fused silica tubing coated with a 1/16 in. O.D. PEEK covering, it is capable of withstanding high pressures. PEEKsil tubing is ideal for capillary HPLC and LC-mass spectrometry (MS). The very smooth internal surface ensures excellent flow characteristics and very low carryover. It is compatible with most organic solvents and is usable in most standard chromatography systems.
Recommended Use
  • Tubing is stiff; not recommended for uses requiring that tubing bends
  • Use precut lengths only; tube cutting after receipt may damage tubing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HPLC, LC/MS
0.007 in. ID; 10 cm length
0.007 in.
0.18 mm
3.937 in.
PEEK, Silica
Capillary Tubing
HPLC, LC/MS
1/16 in.
1.588 mm
10 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ PEEKsil™ Capillary Tubing > 0.007 in. ID; 10cm length