missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE1C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13785-1-AP-20UL
This item is not returnable.
View return policy
Description
Cyclic nucleotide phosphodiesterases (PDEs) catalyze hydrolysis of the cyclic nucleotides cAMP and cGMP to the corresponding nucleoside 5-prime-monophosphates. Mammalian PDEs have been classified into several families based on their biochemical properties. Members of the PDE1 family, such as PDE1C, are calmodulin -dependent PDEs (CaM-PDEs) that are stimulated by a calcium-calmodulin complex (Repaske et al., 1992 ).Specifications
PDE1C | |
Polyclonal | |
Unconjugated | |
PDE1C | |
Cam PDE 1C, hCam 3, Hcam3, PDE1C | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18575, 5137, 81742 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q14123, Q63421, Q64338 | |
PDE1C | |
PDE1C Fusion Protein Ag4798 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |