Learn More
Pararosaniline Solution, Fisher Chemical™
Click to view available options
Quantity:
100 mL
Specifications
Specifications
pH | 2.1 to 2.2 |
Quantity | 100 mL |
Safety and Handling
Product Identifier
- Decolorised Pararosaniline Solution. European Pharmacopoeia Ref: 1062201. (RP 143)
- Danger
- Carcinogenicity Category 1B
- H350-May cause cancer.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to .