Learn More
Cytiva™ Whatman™ Qualitative Filter Paper Grade CF 12
Whatman™ Grade CF 12 qualitative filter paper is ideal for general laboratory applications, such as removing particulate matter, clarifying liquids and separating solids from liquids in qualitative analysis. The Grade CF 12 offers medium retention and flow rate.
Brand:  Cytiva™ 10535097
Product Code. 11915376
  • 492.00 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Qualitative filter paper sheets
  • Grade CF 12
  • Ideal for general filtration applications
  • Medium retention and flow rate
  • Suitable for various industries including pharmaceutical, environmental and food testing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Qualitative filter paper
Qualitative Filter Paper
Cellulose fibers
Grade CF 12
Whatman™
100 Pk.
Sheets 210 x 297 mm
Suitable for various industries including pharmaceutical, environmental and food testing
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva™ Whatman™ Qualitative Filter Paper Grade CF 12 > 210 x 297 mm