Learn More
Papaverine hydrochloride, 99%, Thermo Scientific Chemicals
59.19 € - 446.22 €
Chemical Identifiers
CAS | 61-25-6 |
---|---|
Molecular Formula | C20H21NO4·ClH |
Molecular Weight (g/mol) | 375.85 |
MDL Number | MFCD00012745 |
InChI Key | UOTMYNBWXDUBNX-UHFFFAOYSA-N |
Synonym | papaverine hydrochloride, cardiospan, cardoverina, dispamil, drapavel, forpavin, papalease, papaversan, pavatest, paverolan |
PubChem CID | 6084 |
IUPAC Name | 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxyisoquinoline;hydrochloride |
SMILES | COC1=C(C=C(C=C1)CC2=NC=CC3=CC(=C(C=C32)OC)OC)OC.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10522421
|
Thermo Scientific Acros
207220050 |
5 g | Glass bottle |
59.19 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10606914
|
Thermo Scientific Acros
207220250 |
25 g | Glass bottle |
183.92 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10107843
|
Thermo Scientific
207221000 |
100 g | Glass bottle |
446.22 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
61-25-6 | |
375.85 | |
UOTMYNBWXDUBNX-UHFFFAOYSA-N | |
6084 | |
COC1=C(C=C(C=C1)CC2=NC=CC3=CC(=C(C=C32)OC)OC)OC.Cl |
C20H21NO4·ClH | |
MFCD00012745 | |
papaverine hydrochloride, cardiospan, cardoverina, dispamil, drapavel, forpavin, papalease, papaversan, pavatest, paverolan | |
1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxyisoquinoline;hydrochloride |
Specifications
61-25-6 | |
White to Beige | |
99% | |
MFCD00012745 | |
15, 7122 | |
Solubility in water: freely soluble. Other solubilities: soluble in alcohol and chloroform,practically insoluble in ether | |
COC1=C(C=C(C=C1)CC2=NC=CC3=CC(=C(C=C32)OC)OC)OC.Cl | |
375.85 | |
375.85 | |
Fine Crystalline Powder |
226.0°C | |
Authentic | |
C20H21NO4·ClH | |
21, 222 | |
papaverine hydrochloride, cardiospan, cardoverina, dispamil, drapavel, forpavin, papalease, papaversan, pavatest, paverolan | |
UOTMYNBWXDUBNX-UHFFFAOYSA-N | |
1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxyisoquinoline;hydrochloride | |
6084 | |
99% | |
Papaverine hydrochloride |
Safety and Handling
GHS H Statement
Toxic if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-502-1
- Papaverine hydrochloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.