missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pamoic acid, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
Pamoic acid is used in preparation of repository derivatives of medicinal agents, used in pharmacology to increase the solubility of a drug in water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pamoic acid is used in preparation of repository derivatives of medicinal agents, used in pharmacology to increase the solubility of a drug in water.
Solubility
Soluble in nitrobenzene, pyridine
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Pamoic acid is used in preparation of repository derivatives of medicinal agents, used in pharmacology to increase the solubility of a drug in water.
Solubility
Soluble in nitrobenzene, pyridine
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
Melting Point | >300°C |
Assay Percent Range | 99% |
Beilstein | 901319 |
Merck Index | 14,7005 |
Solubility Information | Soluble in nitrobenzene, pyridine |
Formula Weight | 388.38 |
Percent Purity | 99% |
Quantity | 250 g |
Chemical Name or Material | Pamoic acid |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction