Learn More
Palladium black, 99.9% (metals basis), Thermo Scientific Chemicals
Use as a catalyst in hydrogenation reactions. Other uses include the manufacture of paints, adhesives, fibers and coatings. Basic particles of Palladium black have the tendency to cluster into macroscopic aggregates.
332.66 € - 1564.79 €
Chemical Identifiers
CAS | 7440-05-3 |
---|---|
Molecular Formula | Pd |
Molecular Weight (g/mol) | 106.42 |
MDL Number | MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 |
InChI Key | KDLHZDBZIXYQEI-UHFFFAOYSA-N |
Synonym | black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon |
PubChem CID | 23938 |
ChEBI | CHEBI:33363 |
IUPAC Name | palladium |
SMILES | [Pd] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11386607
|
Thermo Scientific Alfa Aesar
000659.03 |
1 g |
332.66 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11396607
|
Thermo Scientific Alfa Aesar
000659.06 |
5 g |
1564.79 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a catalyst in hydrogenation reactions. Further it finds use in the manufacture of paints, adhesives, fibers and coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-05-3 | |
106.42 | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
23938 | |
palladium |
Pd | |
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 | |
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon | |
CHEBI:33363 | |
[Pd] |
Specifications
Gray to Silver | |
1555°C | |
Powder | |
99.9% (Metals basis) | |
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 | |
Insoluble in water; Soluble in aqua regia and fused alkalies | |
[Pd] | |
106.42 | |
CHEBI:33363 | |
Powder |
7440-05-3 | |
3067°C | |
12.02g/cm3 at 20°C | |
Pd | |
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
palladium | |
23938 | |
Odorless | |
Palladium black |
Safety and Handling
H228: Flammable solid.
missing translation for 'dotInformation' : DOT Class: 4.1, Packaging Group: II
missing translation for 'einecsNumber' : 231-115-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Palladium black
Signal Word
- Danger
Hazard Category
- Flammable solid Category 1
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P370+P378-In case of fire: Use to extinguish.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.