missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium, 20% on activated carbon powder, eggshell, unreduced, nominally 50% water wet, Thermo Scientific Chemicals
Debenzylation
318.00 € - 3740.00 €
Chemical Identifiers
CAS | 7440-05-3 |
---|---|
Molecular Formula | Pd |
Molecular Weight (g/mol) | 106.42 |
MDL Number | MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 |
InChI Key | KDLHZDBZIXYQEI-UHFFFAOYSA-N |
Synonym | black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon |
PubChem CID | 23938 |
ChEBI | CHEBI:33363 |
IUPAC Name | palladium |
SMILES | [Pd] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11314427
|
Thermo Scientific Alfa Aesar
038307.06 |
5 g |
318.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11394417
|
Thermo Scientific Alfa Aesar
038307.14 |
25 g |
1497.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11304427
|
Thermo Scientific Alfa Aesar
038307.22 |
100 g | |||||||
Description
Debenzylation
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-05-3 | |
106.42 | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
23938 | |
palladium |
Pd | |
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 | |
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon | |
CHEBI:33363 | |
[Pd] |
Specifications
7440-05-3 | |
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
palladium | |
23938 | |
Odorless |
Pd | |
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon | |
[Pd] | |
106.42 | |
CHEBI:33363 | |
Palladium, 20% on activated carbon powder, Eggshell, unreduced, nominally 50% water wet |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only