Learn More
p65/RELA Rabbit anti-Human, Mouse, Porcine, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10745-1-AP-20UL
 View more versions of this product
Product Code. 16817333
  • 145.00 € / 20µL
Explore more special offers
Description
Description
This antibody is a rabbit polyclonal antibody raised against residues near the N terminus of human RELA.
The transcription factor NFkB is widely recognized as a critical mediator of immune and inflammatory responses. In most cell types, NFkB is found in the cytoplasm where it is associated with an inhibitory protein known as IkB. An impressive variety of stimuli (tumor necrosis factor, interleukin 1, T-cell activation signals, bacterial endotoxins, viral transforming proteins, certain growth factors and reactive oxygen intermediates) lead to the rapid nuclear assumulation of NFkB by the induced phosphorylation and subsquent degradation of IkB. In the nucleus, NFkB regulates genes encoding cytokines, cytokine receptors, cell adhesion molecules, proteins involved in coagulation and genes involved in cell growth control. Additionally, NFkB is thought to be an important transcriptional regulator for HIV. Growing evidence indicates that the dysregulation of NFkB may be key to a number of diseases including arthritis and other inflammatory diseases, Alzheimer′s disease, atherosclerosis, and cancer.
Specifications
Specifications
p65/RELA
Polyclonal
Unconjugated
RELA
NFKB 3, NFKB p65, NFKB3, p65, RELA, Transcription factor p65
Rabbit
Antigen Affinity Chromatography
RUO
100135665, 19697, 309165, 5970
-20°C
Liquid
Flow Cytometry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry
0.52 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q04206, Q04207
RELA
p65; RELA Fusion Protein Ag1199
20 μL
Primary
Human, Porcine, Rat, Mouse