missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P62/SQSTM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 € - 399.00 €
Specifications
Antigen | P62/SQSTM1 |
---|---|
Concentration | 0.29 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16838474
|
Proteintech
18420-1-AP-20UL |
20 μL |
140.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16828474
|
Proteintech
18420-1-AP-150UL |
150 μL |
399.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The molecular weight of p62 is predicted to be 48/ 38 kDa (depending on the isoform), while western blot analyses using this antibody detects the bands around 45-48 kDa and 60-62 kDa, respectively.
This gene encodes a multifunctional protein that binds ubiquitin and regulates activation of the nuclear factor kappa-B signaling pathway. The protein functions as a scaffolding/adaptor protein in concert with TNF receptor-associated factor 6 to mediate activation of NF-kB in response to upstream signals. Alternatively spliced transcript variants encoding either the same or different isoforms have been identified for this gene. Mutations in this gene result in sporadic and familial Paget disease of bone.Specifications
P62/SQSTM1 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
O08623, Q13501, Q64337 | |
113894, 18412, 8878 | |
P62/SQSTM1 Fusion Protein Ag13131 | |
Primary | |
-20°C | |
SQSTM1 |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P62, sequestosome 1, SQSTM1, Ubiquitin binding protein p62 | |
Sqstm1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |