missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ P2-CST Multiple Gradient Gel Caster, Frosted Notched Glass, 20cm L x 20cm W x 0.18 in. thickness
Cast one to five gels simultaneously with the Fisherbrand™ P2-CST Multiple Gradient Gel Caster, one of the simplest devices to cast gradient gels.
Brand: Fisherbrand™ P2-20FR
This item is not returnable.
View return policy
Description
Cast one to five gels simultaneously with the Fisherbrand™ P2-CST Multiple Gradient Gel Caster, one of the simplest devices to cast gradient gels.
- With an external gradient maker, gels can be cast with ease and great reproducibility
- The design and format of this unit will provide years of service in your lab
Specifications
Frosted notched glass | |
Notched Glass | |
20 cm |
Fisherbrand P2-CST System | |
0.18 in. | |
20 cm |