missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ P1-CST Multiple Gradient Gel Caster, 2 teeth, tooth thickness, 1.5mm
Cast one to five gels simultaneously with Fisherbrand™ P1-CST Multiple Gradient Gel Caster, one of the simplest devices to cast gradient gels.
Brand: Fisherbrand™ P1PREP
This item is not returnable.
View return policy
Description
Cast one to five gels simultaneously
- With an external gradient maker, gels can be cast with ease and great reproducibility
- The design and format of this unit will provide years of service in your lab
Specifications
Prep comb with 2 teeth | |
Comb | |
1.5 mm |
Fisherbrand P1-CST System | |
2 | |
1.5 mm |