missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Xylene, 99%, for HPLC, Thermo Scientific Chemicals
28.04 € - 237.18 €
Chemical Identifiers
CAS | 106-42-3 |
---|---|
Molecular Formula | C8H10 |
Molecular Weight (g/mol) | 106.17 |
MDL Number | MFCD00008556 |
InChI Key | URLKBWYHVLBVBO-UHFFFAOYSA-N |
Synonym | p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene |
PubChem CID | 7809 |
ChEBI | CHEBI:27417 |
IUPAC Name | 1,4-xylene |
SMILES | CC1=CC=C(C)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11984441
|
Thermo Scientific Acros
445681000 |
100 mL | Glass Bottle |
28.04 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11964441
|
Thermo Scientific Acros
445680010 |
1 L | Glass Bottle |
109.55 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11974441
|
Thermo Scientific Acros
445680025 |
2.5 L | Glass Bottle |
237.18 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
106-42-3 | |
106.17 | |
URLKBWYHVLBVBO-UHFFFAOYSA-N | |
7809 | |
1,4-xylene |
C8H10 | |
MFCD00008556 | |
p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene | |
CHEBI:27417 | |
CC1=CC=C(C)C=C1 |
Specifications
13.0°C | |
106-42-3 | |
138.0°C | |
1.4940 to 1.4960 | |
0.866 | |
C6H4(CH3)2 | |
0.648 mPa.s (20°C) | |
Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, ether and most organic, solvents | |
CC1=CC=C(C)C=C1 | |
106.17 | |
CHEBI:27417 | |
99% | |
p-Xylene |
0.01 max. at 380nm, 0.01 max. at 400nm, 0.05 max. at 350nm, 0.1 max. at 320nm, 1 max. at 294nm | |
0.8660g/mL | |
25°C | |
99% | |
C8H10 | |
MFCD00008556 | |
p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene | |
URLKBWYHVLBVBO-UHFFFAOYSA-N | |
1,4-xylene | |
7809 | |
106.17 | |
HPLC |
Safety and Handling
Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- p-Xylene
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
p-Xylene, 99%, for HPLC, Thermo Scientific Chemicals