Learn More
p-Toluoyl chloride, 98%, Thermo Scientific Chemicals
29.55 € - 350.00 €
Chemical Identifiers
CAS | 874-60-2 |
---|---|
Molecular Formula | C8H7ClO |
Molecular Weight (g/mol) | 154.6 |
MDL Number | MFCD00000696 |
InChI Key | NQUVCRCCRXRJCK-UHFFFAOYSA-N |
Synonym | p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4 |
PubChem CID | 13405 |
IUPAC Name | 4-methylbenzoyl chloride |
SMILES | CC1=CC=C(C=C1)C(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10521201
|
Thermo Scientific Acros
147980050 |
5 mL | Glass bottle |
29.55 €
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10666152
|
Thermo Scientific Acros
147982500 |
250 mL | Glass bottle |
68.24 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10108103
|
Thermo Scientific Acros
147980010 |
1 L | Glass bottle |
189.95 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13408378
|
Thermo Scientific Acros
147980025 |
2.5 L | Glass bottle | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
874-60-2 | |
154.6 | |
NQUVCRCCRXRJCK-UHFFFAOYSA-N | |
13405 | |
CC1=CC=C(C=C1)C(=O)Cl |
C8H7ClO | |
MFCD00000696 | |
p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4 | |
4-methylbenzoyl chloride |
Specifications
874-60-2 | |
Colorless to Brown | |
225.0°C to 227.0°C | |
Authentic | |
C8H7ClO | |
1.5525 to 1.5545 | |
09,486 | |
p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4 | |
NQUVCRCCRXRJCK-UHFFFAOYSA-N | |
4-methylbenzoyl chloride | |
13405 | |
98% | |
p-Toluoyl chloride, 98% |
-2.0°C | |
1.1600g/mL | |
82°C | |
98% | |
CH3C6H4COCl | |
MFCD00000696 | |
1.16 | |
Solubility in water: decomposes. | |
CC1=CC=C(C=C1)C(=O)Cl | |
154.6 | |
154.6 | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 212-864-8
missing translation for 'rtecsNumber' : XV1660000
missing translation for 'tsca' : TSCA
- p-Toluoyl chloride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.