Learn More
P-cadherin Rabbit anti-Human, Biotin, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech BIOTIN-13773-100UL
This item is not returnable.
View return policy
Description
The P-cadherin antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human P-cadherin. This antibody recognizes human, mouse, rat antigen. The P-cadherin antibody has been validated for the following applications: IHC analysis. This antibody is biotin.
Specifications
P-cadherin | |
Polyclonal | |
Biotin | |
100 μL | |
1001 | |
IgG |
Immunohistochemistry | |
500 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |