Learn More
Welch™ Chemvac P12Z - 301 Combination Pump
Draws a deep, high-flow vacuum with a powerful Rotary Vane Pump. Welch™ Chemvac P12Z - 301 degasses harmful fumes with a secondary vacuum.
Brand: Welch™ 109031
This item is not returnable.
View return policy
Description
- Comprised of two-stage rotary vane pump P 6 Z and two-stage diaphragm pump MPC 301 Z
- Suitable for freeze dry applications
- Both pumps are electrical and vacuum-connected
- Rotary vane pump includes an outtake with a separator for the collection of condensates.
- Supplied completed with solenoid valve block
Specifications
115/230 V 50/60 Hz | |
P 12 Z - 301 | |
420 mm | |
Chemvac Combination Pump System | |
115/230 V |
345 mm | |
50/60 Hz | |
590 mm | |
2 x 10-3 mbar |