missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxalic acid, Thermo Scientific Chemicals
38.89 € - 74.07 €
Chemical Identifiers
CAS | 144-62-7 |
---|---|
Molecular Formula | C2H2O4 |
Molecular Weight (g/mol) | 90.034 |
MDL Number | MFCD00002573 |
InChI Key | MUBZPKHOEPUJKR-UHFFFAOYSA-N |
Synonym | ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico |
PubChem CID | 971 |
ChEBI | CHEBI:16995 |
IUPAC Name | oxalic acid |
SMILES | C(=O)(C(=O)O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10397190
|
Thermo Scientific
124280010 |
1 L | Plastic bottle |
38.89 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10331374
|
Thermo Scientific Acros
124280025 |
2.5 L | Plastic bottle |
74.07 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Volumetric standard solutionChemical Identifiers
144-62-7 | |
90.034 | |
MUBZPKHOEPUJKR-UHFFFAOYSA-N | |
971 | |
oxalic acid |
C2H2O4 | |
MFCD00002573 | |
ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico | |
CHEBI:16995 | |
C(=O)(C(=O)O)O |
Specifications
144-62-7 | |
C2H2O4 | |
MFCD00002573 | |
15, 7010 | |
Solubility in water: soluble in water. | |
C(=O)(C(=O)O)O | |
oxalic acid | |
971 | |
90.04 | |
Liquid | |
Oxalic acid |
Colorless | |
HO2CCO2H | |
02, 502 | |
ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico | |
MUBZPKHOEPUJKR-UHFFFAOYSA-N | |
0.0998 to 0.1002N (20°C) | |
90.034 | |
CHEBI:16995 | |
0.9900g/mL | |
0.99 |
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Oxalic acid, Thermo Scientific Chemicals