missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System
Run 10 to 80 samples on one gel with this wide-gel electrophoresis system.
Brand: Thermo Scientific™ D2
This item is not returnable.
View return policy
Description
Run 10 to 80 samples on one gel with the Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System.
- Wide gel size allows 15 or 30 samples to be loaded per row with a micro well comb.
- Using special combs and a multichannel pipetter, samples may be loaded directly from a 96-well plate, eight or 12 at a time.
- System is excellent for screening PCR samples or routine DNA fragment analysis.
- Small footprint saves space on lab bench.
- Incorporates Thermo Scientific™ EasyCast™ casting design with external caster—gel tray is fitted with gasket, allowing it to fit snugly into caster and providing a leakproof seal without tape.
- Electrode placement allows gel to be run quickly without compromising resolution.
Includes:
- Buffer chamber
- Thermo Scientific™ SuperSafe™ lid with attached power supply leads
- EasyCast gasketed UVT gel tray
- External multiple gel caster
- Four combs (two 30-well and two 40-well, 1.5mm thick)
Specifications
CE marked | |
Owl | |
10 to 80 | |
7.38 x 7.75 x 4 in. (19 x 20 x 10 cm) | |
19 cm | |
7.75 in. | |
20 cm | |
4 in. | |
150 V |
D2 Wide-Gel | |
Wide | |
600 mL | |
7.38 in. | |
14.4 cm | |
6.5 | |
10.2 cm | |
10 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System > D2 Wide-Gel Complete System without Buffer Exchange Ports
Spot an opportunity for improvement?Share a Content Correction