missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osteocalcin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23418-1-AP-150UL
This item is not returnable.
View return policy
Description
Osteocalcin belongs to the osteocalcin/matrix Gla-protein family and constitutes 1-2% of the total bone protein. It binds strongly to apatite and calcium. Gamma-carboxyglutamate residues are formed by vitamin K dependent carboxylation. These residues are essential for the binding of calcium.Specifications
Osteocalcin | |
Polyclonal | |
Unconjugated | |
BGLAP | |
BGLAP, Osteocalcin, BGP, Bone Gla protein, OC, OCN, Osteocalcin, PMF1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12096, 25295, 632 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P02818, P04640, P86546 | |
BGLAP | |
Osteocalcin Fusion Protein Ag20065 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction