Learn More
Osteocalcin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 23418-1-AP-20UL
 View more versions of this product
Product Code. 16802345
  • 145.00 € / 20µL
Explore more special offers
Description
Description
Osteocalcin belongs to the osteocalcin/matrix Gla-protein family and constitutes 1-2% of the total bone protein. It binds strongly to apatite and calcium. Gamma-carboxyglutamate residues are formed by vitamin K dependent carboxylation. These residues are essential for the binding of calcium.
Specifications
Specifications
Osteocalcin
Polyclonal
Unconjugated
BGLAP
BGLAP, Osteocalcin, BGP, Bone Gla protein, OC, OCN, Osteocalcin, PMF1
Rabbit
Antigen Affinity Chromatography
RUO
12096, 25295, 632
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin)
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P02818, P04640, P86546
BGLAP
Osteocalcin Fusion Protein Ag20065
20 μL
Primary
Rat, Mouse, Human