missing translation for 'onlineSavingsMsg'
Learn More

Osmium powder, -20 mesh, 99.95% (metals basis), Thermo Scientific Chemicals

221.10 € - 733.65 €

Chemical Identifiers

CAS 7440-04-2
PubChem CID 23937
ChEBI CHEBI:30687
IUPAC Name osmium
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11369197
View Documents Promotion Details
Thermo Scientific Alfa Aesar
039998.03
1 g
221.10 €
1g
Please to purchase this item. Need a web account? Register with us today!
11359197
View Documents Promotion Details
Thermo Scientific Alfa Aesar
039998.06
5 g
733.65 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-04-2
23937
CHEBI:30687
osmium

Specifications

7440-04-2
Os
ion, metallic, osmium, elemental, unii-2e7m255opy, os4, osmio, hydrido, hydride, atom, powder
[Os]
190.23
CHEBI:30687
Osmium powder
99.95% (metals basis)
MFCD00011147
SYQBFIAQOQZEGI-UHFFFAOYSA-N
osmium
23937
-20 Mesh
Videos
Safety and Handling

Safety and Handling

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H318-H335

missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: II

missing translation for 'einecsNumber' : 231-114-0

missing translation for 'rtecsNumber' : RN1100000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only