missing translation for 'onlineSavingsMsg'
Learn More

Osmium(IV) oxide, Os 83% min, Thermo Scientific Chemicals

126.63 € - 1434.14 €

Chemical Identifiers

CAS 12036-02-1
Molecular Formula O2Os
Molecular Weight (g/mol) 222.228
MDL Number MFCD00011150
InChI Key XSXHWVKGUXMUQE-UHFFFAOYSA-N
Synonym osmium iv oxide, osmium dioxide, osmium oxide oso2 6ci,7ci,8ci,9ci, osmium iv-oxid, acmc-1btxg, osmium iv oxide, os
PubChem CID 187574
IUPAC Name dioxoosmium
SMILES O=[Os]=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11388298
SDS View Documents
Thermo Scientific Alfa Aesar
039497.01
0.25 g
126.63 €
0.25g
Please to purchase this item. Need a web account? Register with us today!
11378298
SDS View Documents
Thermo Scientific Alfa Aesar
039497.03
1 g
363.81 €
1g
Please to purchase this item. Need a web account? Register with us today!
11368298
SDS View Documents
Thermo Scientific Alfa Aesar
039497.06
5 g
1434.14 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The osmium oxide/hexacyanoruthenate films modified electrodes plays an important role for the electrocatalytic reaction of catecholamines and sulfur oxoanions. Used as catalyst. Metallic osmium or an osmium oxide-ruthenium cyanide film modified graphite electrodes plays an essential role for the determination of biogenic amines from electrocatalytic responses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Osmium(IV) oxide, Os 83% min, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.