Learn More
Osmium(III) chloride hydrate, Thermo Scientific Chemicals
154.77 € - 518.58 €
Chemical Identifiers
CAS | 14996-60-2 |
---|---|
Molecular Formula | Cl3H2OOs |
Molecular Weight (g/mol) | 314.60 |
MDL Number | MFCD00149815 |
InChI Key | KHHOLOMEVBPRQY-UHFFFAOYSA-K |
Synonym | osmium iii chloride hydrate, osmium chloride oscl3 , hydrate 8ci,9ci, trichloroosmium hydrate, cl3os.h2o, trichloroosmium-water 1/1 |
PubChem CID | 16211485 |
SMILES | O.Cl[Os](Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10450351
|
Thermo Scientific Acros
318705000 |
500 mg | Glass bottle |
154.77 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10721555
|
Thermo Scientific Acros
318700025 |
2.5 g | Glass bottle |
518.58 €
2.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14996-60-2 | |
314.60 | |
KHHOLOMEVBPRQY-UHFFFAOYSA-K | |
16211485 |
Cl3H2OOs | |
MFCD00149815 | |
osmium iii chloride hydrate, osmium chloride oscl3 , hydrate 8ci,9ci, trichloroosmium hydrate, cl3os.h2o, trichloroosmium-water 1/1 | |
O.Cl[Os](Cl)Cl |
Specifications
>500.0°C | |
Black to Red | |
Cl3H2OOs | |
MFCD00149815 | |
Solubility in water: soluble | |
O.Cl[Os](Cl)Cl | |
314.60 | |
296.56 |
14996-60-2 | |
Crystals or Powder | |
OsCl3·xH2O | |
osmium iii chloride hydrate, osmium chloride oscl3 , hydrate 8ci,9ci, trichloroosmium hydrate, cl3os.h2o, trichloroosmium-water 1/1 | |
KHHOLOMEVBPRQY-UHFFFAOYSA-K | |
trichloroosmium;hydrate | |
16211485 | |
Osmium(III) chloride hydrate |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for s
GHS Signal Word: Danger
RUO – Research Use Only