Learn More
Osmium 99.9%, Thermo Scientific Chemicals
158.00 € - 540.00 €
Chemical Identifiers
CAS | 7440-04-2 |
---|---|
Molecular Formula | Os |
Molecular Weight (g/mol) | 190.23 |
MDL Number | MFCD00011147 |
InChI Key | SYQBFIAQOQZEGI-UHFFFAOYSA-N |
Synonym | ion, metallic, osmium, elemental, unii-2e7m255opy, os4, osmio, hydrido, hydride, atom, powder |
PubChem CID | 23937 |
ChEBI | CHEBI:30687 |
IUPAC Name | osmium |
SMILES | [Os] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10531844
|
Thermo Scientific Acros
385160010 |
1 g | Glass bottle |
158.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10045483
|
Thermo Scientific Acros
385160050 |
5 g | Glass bottle |
540.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisChemical Identifiers
7440-04-2 | |
190.23 | |
SYQBFIAQOQZEGI-UHFFFAOYSA-N | |
23937 | |
osmium |
Os | |
MFCD00011147 | |
ion, metallic, osmium, elemental, unii-2e7m255opy, os4, osmio, hydrido, hydride, atom, powder | |
CHEBI:30687 | |
[Os] |
Specifications
Black | |
3045.0°C | |
99.90% | |
Os | |
15, 6988 | |
Solubility in water: insoluble. | |
[Os] | |
190.23 | |
CHEBI:30687 | |
99.9% | |
Powder |
7440-04-2 | |
5027.0°C | |
Os | |
MFCD00011147 | |
ion, metallic, osmium, elemental, unii-2e7m255opy, os4, osmio, hydrido, hydride, atom, powder | |
SYQBFIAQOQZEGI-UHFFFAOYSA-N | |
osmium | |
23937 | |
190.2 | |
Trace Metal Basis | |
Osmium |
Safety and Handling
GHS H Statement:
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only