missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Orthophosphoric Acid, 85+%, for Electronic Use, Fisher Chemical
Click to view available options
Quantity:
2.5 L
Packaging:
Plastic Coated Glass bottle
Specifications
Specifications
Boiling Point | 158°C |
Melting Point | 21°C |
CAS | 7664-38-2 |
pH | 2 |
Physical Form | Liquid |
Quantity | 2.5 L |
Molecular Formula | H3O4P |
MDL Number | 11340 |
Synonym | orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico |
InChI Key | NBIIXXVUZAFLBC-UHFFFAOYSA-N |
Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- Phosphoric acid, 85+% solution in water
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Orthophosphoric acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'