missing translation for 'onlineSavingsMsg'
Learn More

ORP test solution, 200-275 mV, Thermo Scientific Chemicals

67.94 € - 92.56 €

Specifications

Chemical Name or Material ORP test solution
Packaging Plastic-coated glass bottle
MDL Number MFCD01867801
Physical Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15451787
View Documents Promotion Details
Thermo Scientific Alfa Aesar
042131.AK
250 mL
67.94 €
250mL
Please to purchase this item. Need a web account? Register with us today!
15461787
View Documents Promotion Details
Thermo Scientific Alfa Aesar
042131.AP
500 mL
92.56 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Test solution for ORP electrodes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

ORP test solution
Plastic-coated glass bottle
MFCD01867801
Liquid
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only