Learn More
Thermo Scientific™ Orion™ Electrode Adapter Cables
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
Brand:  Thermo Scientific™ 090036
 View more versions of this product
Product Code. 11986079
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
  • For use with meters that have BNC, US standard, certain LEMO, British, E DIN, Radiometer, 2mm reference pins or just a stripped lead inputs
  • Electrodes must have a US standard, screw cap, Karl Fisher, or BNC connectors
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BNC
Meters with 2 mm Reference Pins Connectors, Meters with BNC Connectors, Meters with British Connectors, Meters with Certain LEMO Connectors, Meters with E DIN Connectors, Meters with Radiometer Connectors, Meters with Stripped Lead Inputs Connectors, Meters with US Standard Connectors
F LEMO (Metrohm)
F LEMO (Metrohm); BNC; BNC to F LEMO Adapter
Electrode Adapter Cable