Learn More
Orion™ 3-Star Plus pH Portable Meter
Confidently gather samples for the most demanding of field applications with the Thermo Scientific™ Orion™ 3-Star Plus pH Portable Meter.
Brand: Thermo Scientific Orion 1212501
Description
Confidently gather samples for the most demanding of field applications with the waterproof, battery-operated Thermo Scientific™ Orion™ 3-Star Plus pH Portable Meter, which features highly accurate calibration and measurement.
Easy to operate in any mode:
- Bright, backlit LCD display with ready indication
- Balanced comfortable handling
- Delivers over 2000 hours of continuous operation
- Simultaneous display of pH or mV and temperature
- Convenient built-in Quick-Start Guide with calibration instructions
- Stores, displays and prints up to 10 methods with individual method password protection
- Automatic or manual temperature compensation
- Auto-read/hold function
- IP67 waterproof
Extra features in every mode:
- pH: automatic or manual calibration; one to five calibration points; auto-buffer-recognition of NIST™/US and DIN buffers
- mV/ORP: EH autocalibration to standard hydrogen electrode
Increased accuracy:
- Improved stability algorithm that improves calibration and measurement accuracy
- Thermo Scientific™ SMART STABILITY™ and SMART AVERAGING™ functions that optimize accuracy, precision and response time
- ATC calibration
- Advanced self-tests on meter operation, self-diagnostics
Complete documentation:
- Full complement of GLP-related features
- 1000 point datalogger, downloadable to printer or computer via RS-232 connection
- Average slope and slope of segment printed with measurement
Includes:
Meter Only 1212000: meter; batteries and instructions.
Meter Kit/pH 1212001: meter; pH/ATC Triode™ 9107BNMD; 10 x 15mL pouches each of pH 4.01/7.00/10.01 buffers; 5 x 60mL pH electrode storage solution; beaker; electrode storage chamber; hard field case; batteries and instructions.
Meter Kit/pH 1212002: meter; pH/ATC NoCal™ electrode 5107BNMD; 10 x 15mL pouches each of pH 4.01/7.00/10.01 buffers; 5 x 60mL pH electrode storage solution; beaker; hard field case; batteries and instructions.
Meter Kit/pH 1212005: meter; pH/ATC Triode 8107BNMD; 10 x 15mL pouches each of pH 4.01/7.00/10.01 buffers; 5 x 60mL pH electrode storage solution; beaker, electrode storage chamber, soft field case; batteries and instructions.
Meter Kit/pH 1212503: meter; ORP/ATC Triode 9179BNMD; 5 x 60mL ORP Standard; meter impact jacket with electrode storage chamber, soft field case; batteries and instructions.
Meter Kit/pH 1212101 package includes meter; pH/ATC Triode 9107BNMD; batteries and instructions.
Recommended Accessories and Replacements
- Triode pH/ATC Electrode 9107BNMD
- pH Electrode Storage Solution 910060
- Meter Impact Jacket 1210002
- pH Buffer perpHect™ Individual Pouches 910410, 911010, 910710
- RS-232 to USB Adapter 1010005
- Star Plus Navigator 21 Software 1010017; complete software for 21 CFR Part II Compliance
Specifications
Meter Kit 1212501 | |
±0.2 or ±0.05%, whichever is greater | |
RS-232, Stereo Jack, Optional USB Interface Cable | |
0.001, 0.01, 0.1 | |
0.1mV | |
0.1°C up to 99.9°C, 1.0°C over 99.9°C | |
Waterproof BNC for pH/ORP electrodes; waterproof 8-Pin Mini-DIN Locking for ATC probe; Pin tip for separate reference electrode | |
IP-67 waterproof | |
SMART STABILITY™ and SMART AVERAGING™: Resolution dependent/automatic | |
4 x AA |
±0.1°C, ±1 digit | |
±0.002 | |
-2.000 to 19.999 | |
±1999.9 | |
-5°C to +105°C | |
1000 with Date and Time Stamp | |
21 CFR Part II Compliance with optional software. CE, CSA, UL and FCC Class A limits approved. | |
5 to 85%, non-condensing | |
5°C to 45°C (ambient) |