missing translation for 'onlineSavingsMsg'
Learn More

OPTIZYME™ AluI, Fisher BioReagents™

Brand:  Fisher BioReagents BP8015-1

Product Code. 11904782

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

5'...A G^C T...3'
3'...T C^G A...5'

Supplied with: 10X OPTIZYME Buffer 4
Conditions for 100% Activity:

  • 1X OPTIZYME Buffer 4: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/ml BSA
  • Incubate at 37°C
Enzyme Activity in OPTIZYME buffers:
  • Buffer 1: 50 - 100%
  • Buffer 2: 0 - 20%
  • Buffer 3: 0 - 20%
  • Buffer 4: 100%
  • Buffer 5: 0 - 20%
Storage Buffer:
  • 10mM Tris-HCl (pH 7.5 at 25°C), 50mM KCl, 1 mM DTT, 0.1mM EDTA, 0.15% Triton X-100, 0.2mg/ml BSA and 50% (v/v) glycerol
  • Ligation and Re-cleavage:
  • More than 95% of the DNA fragments can be ligated and re-cut after a 50-fold over-digestion with AluI
  • Methylation Effects:
  • Dam: Never overlaps - no effect
  • Dcm: Never overlaps - no effect
  • CpG: Never overlaps - no effect
  • EcoKI: Never overlaps - no effect
  • EcoBI: May overlap - blocked
    • 5'...TGm6AGCT(N)5 TGCT...3'
  • 3'...AC TCGA(N)5 m6ACGA...5'
  • (Cleavage is blocked)
  • TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    10 U/μL
    37°C
    >95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with AluI
    Keep container tightly closed
    AG.CT
    Liquid
    25 to 50% Water, >50% Glycerin
    7.4
    10mM Tris HCl (pH 7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.15% Triton™ X-100, 0.2mg/mL BSA, 50% Glycerol
    600 U
    Alul
    Product Suggestions

    Product Suggestions

    Videos
    Safety and Handling

    Safety and Handling

    Product Identifier
    • OPTIZYME

    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers