Learn More
OPRM1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27625-1-AP-20UL
This item is not returnable.
View return policy
Description
The OPRM1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human OPRM1. This antibody recognizes human, mouse, rat antigen. The OPRM1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
OPRM1 | |
Polyclonal | |
Unconjugated | |
20 μL | |
4988 | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
267 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |