Learn More
OPLAH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21127-1-AP-150UL
 View more versions of this product
Product Code. 16800735
  • 410.00 € / 150µL
Explore more special offers
Description
Description
OPLAH catalyzes the cleavage of 5-oxo-L-proline to form L-glutamate coupled to the hydrolysis of ATP to ADP and inorganic phosphate. ATP + 5-oxo-L-proline + 2 H2O = ADP + phosphate + L-glutamate.
Specifications
Specifications
OPLAH
Polyclonal
Unconjugated
OPLAH
5 Opase, 5 oxo L prolinase, 5 oxoprolinase, OPLA, OPLAH, Pyroglutamase
Rabbit
Antigen Affinity Chromatography
RUO
116684, 26873, 75475
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O14841, P97608, Q8K010
Oplah
OPLAH Fusion Protein Ag15523
150 μL
Primary
Human, Mouse, Rat