missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diba Omnifit™ BenchMark™ Replacement O-rings for Column Assemblies
Choose from FKM or FFKM O-rings
POA
Specifications
Type | O-ring |
---|---|
Color | Black |
For Use With (Application) | Chromatography |
For Use With | Omnifit Benchmark Chromatography Columns |
Material | FKM |
Description
Use for quick-disconnect fittings, valves, pumps, and other fluid handling products.Specifications
O-ring | |
Chromatography | |
FKM |
Black | |
Omnifit Benchmark Chromatography Columns |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title